3-Chloro-4'-methoxypropiophenone 95%
Lieferant: Apollo Scientific
Synonyme:
3-Chloro-1-(4-methoxyphenyl)propan-1-one
OR912485-5G
APOSOR912485-5GEA
81.2
EUR
APOSOR912485-5G
3-Chloro-4'-methoxypropiophenone 95%
3-Chloro-4'-methoxypropiophenone
Formel:
C₁₀H₁₁ClO₂ Molecular Weight: 198,65 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236602 CAS-Nummer: 35999-20-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...