(5-Thien-2-ylisoxazol-3-yl)methylamine 95%

Lieferant: Apollo Scientific
Gefahr

OR9125-10MG
APOSOR9125-10MGEA 21.9 EUR
APOSOR9125-10MG
(5-Thien-2-ylisoxazol-3-yl)methylamine 95%
(5-Thien-2-ylisoxazol-3-yl)methylamine
Formel: C₈H₈N₂OS
Molecular Weight: 180,227 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08271879
CAS-Nummer: 852180-45-1
UN: 3259
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR