2,3,4,5-Tetrahydro-1H-benzo[b]azepine 95%

Lieferant: Apollo Scientific

OR912503-5G
APOSOR912503-5GEA 98.6 EUR
APOSOR912503-5G
2,3,4,5-Tetrahydro-1H-benzo[b]azepine 95%
2,3,4,5-Tetrahydro-1H-benzo[b]azepine
Formel: C₁₀H₁₃N
Molecular Weight: 147,2200 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00272363
CAS-Nummer: 1701-57-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR