3-(((tert-Butoxycarbonyl)amino)methyl)benzoic acid 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(BOC-Aminomethyl)benzoic acid, 3-[[(tert-butoxycarbonyl)amino]methyl]benzoic acid, N-Boc-3-Aminomethyl-benzoicacid
OR912508-5G
APOSOR912508-5GEA
66.7
EUR
APOSOR912508-5G
3-(((tert-Butoxycarbonyl)amino)methyl)benzoic acid 96%
3-(((tert-Butoxycarbonyl)amino)methyl)benzoic acid
Formel:
C₁₃H₁₇NO₄ Schmelzpunkt: 126…127 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00273426 CAS-Nummer: 117445-22-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...