2-Chlor-4,6-didimethoxypyrimidin 98%

Lieferant: Apollo Scientific

OR912511-100G
APOSOR912511-100GEA 82.8 EUR
APOSOR912511-100G
2-Chlor-4,6-didimethoxypyrimidin 98%
2-Chlor-4,6-didimethoxypyrimidin
Formel: C₆H₇ClN₂O₂
Molecular Weight: 174,58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00274530
CAS-Nummer: 13223-25-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR