trans-4-tert-Butylcyclohexancarbonsäure 95%
Lieferant: Apollo Scientific
Synonyme:
trans-4-(tert-Butyl)cyclohexanecarboxylic acid
OR912532-5G
APOSOR912532-5GEA
115
EUR
APOSOR912532-5G
trans-4-tert-Butylcyclohexancarbonsäure 95%
trans-4-tert-Butylcyclohexancarbonsäure
Formel:
C₁₁H₂₀O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00466237 CAS-Nummer: 943-29-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...