3-Methyl-1H-indazol 98%

Lieferant: Apollo Scientific

OR912540-5G
APOSOR912540-5GEA 66.7 EUR
APOSOR912540-5G
3-Methyl-1H-indazol 98%
3-Methyl-1H-indazole
Formel: C₈H₈N₂
Molecular Weight: 132.1600 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00601361
CAS-Nummer: 3176-62-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR