(2R)-2-(Isopropylamino)-2-phenylethanol 95%

Lieferant: Apollo Scientific

Synonyme: (R)-2-(Isopropylamino)-2-phenylethanol

OR912554-1G
APOSOR912554-1GEA 314 EUR
APOSOR912554-1G
(2R)-2-(Isopropylamino)-2-phenylethanol 95%
(R)-2-Isopropylamino-2-phenylethanol
Formel: C₁₁H₁₇NO
Molecular Weight: 179.26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00671548
CAS-Nummer: 112211-92-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR