2-Pyrazinecarbonyl chloride 95%

Lieferant: Apollo Scientific

OR912568-1G
APOSOR912568-1GEA 398 EUR
APOSOR912568-1G
2-Pyrazinecarbonyl chloride 95%
2-Pyrazinecarbonyl chloride
Formel: C₅H₃ClN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00792886
CAS-Nummer: 19847-10-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR