4,5,6,7-Tetrahydro-1H-benzoimidazole 95%

Lieferant: Apollo Scientific

Synonyme: 4,5,6,7-Tetrahydro-1H-benzo[d]imidazole

OR912584-5G
APOSOR912584-5GEA 155 EUR
APOSOR912584-5G
4,5,6,7-Tetrahydro-1H-benzoimidazole 95%
4,5,6,7-Tetrahydro-1H-benzoimidazole
Formel: C₇H₁₀N₂
Molecular Weight: 122.168 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00829762
CAS-Nummer: 3752-24-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR