6-Methylsalicylsäure Ethylester 97%

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-Hydroxy-6-methylbenzoate, 2-Hydroxy-6-methylbenzoic Acid Ethyl Ester, 6-Methylsalicylic Acid Ethyl Ester

OR912635-25G
APOSOR912635-25GEA 127 EUR
APOSOR912635-25G
6-Methylsalicylsäure Ethylester 97%
Ethyl-6-methylsalicylate
Formel: C₁₀H₁₂O₃
Molecular Weight: 180.20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01194299
CAS-Nummer: 6555-40-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR