3-Methoxy-2-nitro-6-picoline 95%

Lieferant: Apollo Scientific

OR912641-1G
APOSOR912641-1GEA 806 EUR
APOSOR912641-1G
3-Methoxy-2-nitro-6-picoline 95%
3-Methoxy-2-nitro-6-picoline
Formel: C₇H₈N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01313012
CAS-Nummer: 24015-98-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR