(S)-2-Hydroxybutanoic acid 96%

Lieferant: Apollo Scientific

OR912658-10G OR912658-1G OR912658-5G
APOSOR912658-10GEA 369 EUR
APOSOR912658-10G APOSOR912658-1G APOSOR912658-5G
(S)-2-Hydroxybutanoic acid 96%
(S)-2-Hydroxybutanoic acid
Formel: C₄H₈O₃
Molecular Weight: 104.1045 g/mol
Schmelzpunkt: 50…54 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01318568
CAS-Nummer: 3347-90-8

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR