Methyl 2-methoxy-3-nitrobenzoate 98%

Lieferant: Apollo Scientific
Achtung

OR912665-1G
APOSOR912665-1GEA 466 EUR
APOSOR912665-1G
Methyl 2-methoxy-3-nitrobenzoate 98%
Methyl 2-methoxy-3-nitrobenzoate
Formel: C₉H₉NO₅
Molecular Weight: 211,171 g/mol
Schmelzpunkt: 48…52 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11045625
CAS-Nummer: 90564-26-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR