(S)-5-(Bromomethyl)-2-pyrrolidinone 96%

Lieferant: Apollo Scientific

Synonyme: (S)-5-Bromomethyl-2-pyrrolidinone

OR912689-5G
APOSOR912689-5GEA 184 EUR
APOSOR912689-5G
(S)-5-(Bromomethyl)-2-pyrrolidinone 96%
(S)-5-(Bromomethyl)-2-pyrrolidinone
Formel: C₅H₈BrNO
Molecular Weight: 178.027 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01632456
CAS-Nummer: 72479-05-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR