(2-Cyanophenyl)acetic acid 98%
Lieferant: Apollo Scientific
OR912691-5G
APOSOR912691-5GEA
60.9
EUR
APOSOR912691-5G
(2-Cyanophenyl)acetic acid 98%
(2-Cyanophenyl)acetic acid
Formel:
C₉H₇NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01646238 CAS-Nummer: 18698-99-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...