4-(Aminomethyl)-2,5-dimethyl-1,3-oxazole
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(2,5-Dimethyl-1,3-oxazol-4-yl)methylamine
OR9127-500MG
APOSOR9127-500MGEA
725
EUR
APOSOR9127-500MG
4-(Aminomethyl)-2,5-dimethyl-1,3-oxazole
4-(Aminomethyl)-2,5-dimethyl-1,3-oxazole
Formel:
C₆H₁₀N₂O Molecular Weight: 126,156 g/mol Siedepunkt: 36…38 °C (0,1 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD08271883 CAS-Nummer: 859850-62-7 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...