3-Piperidin-4-ylpropan-1-ol 96%
Lieferant: Apollo Scientific
OR912733-5G
APOSOR912733-5GEA
119
EUR
APOSOR912733-5G
3-Piperidin-4-ylpropan-1-ol 96%
3-Piperidin-4-ylpropan-1-ol
Formel:
C₈H₁₇NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD02178380 CAS-Nummer: 7037-49-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...