5-Methylindole-3-carboxylic acid methyl ester 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 5-methylindole-3-carboxylate
OR912748-5G
APOSOR912748-5GEA
168
EUR
APOSOR912748-5G
5-Methylindole-3-carboxylic acid methyl ester 95%
5-Methylindole-3-carboxylic acid methyl ester
Formel:
C₁₁H₁₁NO₂ Molecular Weight: 189.211 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02179412 CAS-Nummer: 227960-12-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...