4-Ethyl-6-hydroxy-2-methylpyrimidine 95%

Lieferant: Apollo Scientific

OR912842-1G OR912842-250MG OR912842-5G
APOSOR912842-1GEA 0 EUR
APOSOR912842-1G APOSOR912842-250MG APOSOR912842-5G
4-Ethyl-6-hydroxy-2-methylpyrimidine 95%
4-Ethyl-6-hydroxy-2-methylpyrimidine
Formel: C₇H₁₀N₂O
Molecular Weight: 138.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03265397
CAS-Nummer: 52421-75-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR