4-(Propylamino)benzonitrile 98%
Lieferant: Apollo Scientific
OR912846-10G
OR912846-1G
OR912846-5G
APOSOR912846-10GEA
502
EUR
APOSOR912846-10G
APOSOR912846-1G
APOSOR912846-5G
4-(Propylamino)benzonitrile 98%
4-(Propylamino)benzonitrile
Formel:
C₁₀H₁₂N₂ Molecular Weight: 160.216 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11135495 CAS-Nummer: 4714-64-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...