Methyl-3-oxo-3-(2-thienyl)propanoate 95%
Lieferant: Apollo Scientific
Synonyme:
3-Oxo-3-thiophen-2-yl-propionic acid methyl ester, Methyl 3-oxo-3-(thiophen-2-yl)propanoate
OR912862-1G
APOSOR912862-1GEA
458
EUR
APOSOR912862-1G
Methyl-3-oxo-3-(2-thienyl)propanoate 95%
Methyl-3-oxo-3-(2-thienyl)propanoate
Formel:
C₈H₈O₃S Molecular Weight: 184,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03424761 CAS-Nummer: 134568-16-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...