2-Amino-3-chlorbenzonitril 98%

Lieferant: Apollo Scientific

OR912867-25G
APOSOR912867-25GEA 192 EUR
APOSOR912867-25G
2-Amino-3-chlorbenzonitril 98%
2-Amino-3-chlorbenzonitril
Formel: C₇H₅ClN₂
Molecular Weight: 152.58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03425881
CAS-Nummer: 53312-77-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR