Methyl 3-(3-hydroxyphenyl)propionate 97%

Lieferant: Apollo Scientific

OR912911-1G
APOSOR912911-1GEA 77 EUR
APOSOR912911-1G
Methyl 3-(3-hydroxyphenyl)propionate 97%
Methyl 3-(3-hydroxyphenyl)propionate
Formel: C₁₀H₁₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD03788460
CAS-Nummer: 61389-68-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR