5-Ethyl-2,3-dimethylpyrazin 95%
Lieferant: Apollo Scientific
Synonyme:
2,3-Dimethyl-5-ethylpyrazine
OR912937-5G
APOSOR912937-5GEA
59.6
EUR
APOSOR912937-5G
5-Ethyl-2,3-dimethylpyrazin 95%
5-Ethyl-2,3-dimethylpyrazin
Formel:
C₈H₁₂N₂ Molecular Weight: 136.1900 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04038059 CAS-Nummer: 15707-34-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...