[2-(Aminomethyl)phenyl]acetic acid 97%

Lieferant: Apollo Scientific

Synonyme: [2-(Aminomethyl)phenyl]acetic Acid, 2-Aminomethylphenylacetic acid, 2-(2-(Aminomethyl)phenyl)acetic acid

OR912943-1G
APOSOR912943-1GEA 0 EUR
APOSOR912943-1G
[2-(Aminomethyl)phenyl]acetic acid 97%
[2-(Aminomethyl)phenyl]acetic acid
Formel: C₉H₁₁NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04038440
CAS-Nummer: 40851-65-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR