3-Methanesulphonyloxymethylpyrrolidine-1-carboxylic acid tert-butyl ester 97%

Lieferant: Apollo Scientific

Synonyme: 1-Boc-3-[(Methanesulphonyloxy)methyl]pyrrolidine

OR912949-1G
APOSOR912949-1GEA 261 EUR
APOSOR912949-1G
3-Methanesulphonyloxymethylpyrrolidine-1-carboxylic acid tert-butyl ester 97%
3-Methanesulphonyloxymethylpyrrolidine-1-carboxylic acid tert-butyl ester
Formel: C₁₁H₂₁NO₅S
Molecular Weight: 279.36 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04038482
CAS-Nummer: 141699-56-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR