Indan-5-carboxylic acid 95%

Lieferant: Apollo Scientific

Synonyme: Indane-5-carboxylic Acid

OR912972-1G
APOSOR912972-1GEA 148 EUR
APOSOR912972-1G
Indan-5-carboxylic acid 95%
Indan-5-carboxylic acid
Formel: C₁₀H₁₀O₂
Molecular Weight: 162.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04038659
CAS-Nummer: 65898-38-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR