Methyl (3-cyanophenyl)acetate 96%

Lieferant: Apollo Scientific

Synonyme: Methyl 2-(3-cyanophenyl)acetate

OR912976-1G
APOSOR912976-1GEA 142 EUR
APOSOR912976-1G
Methyl (3-cyanophenyl)acetate 96%
Methyl (3-cyanophenyl)acetate
Formel: C₁₀H₉NO₂
Molecular Weight: 175,184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04038679
CAS-Nummer: 52798-00-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR