Ethyl 2-aminopyrimidine-5-carboxylate 95%

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-aminopyrimidine-5-carboxylate, 2-Amino-pyrimidine-5-carboxylic acid ethyl ester

OR912994-5G
APOSOR912994-5GEA 134 EUR
APOSOR912994-5G
Ethyl 2-aminopyrimidine-5-carboxylate 95%
Ethyl-2-aminopyrimidine-5-carboxylate
Formel: C₇H₉N₃O₂
Molecular Weight: 167.1700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04114100
CAS-Nummer: 57401-76-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR