3-Chloro-5-nitropyridine 95%

Lieferant: Apollo Scientific

OR912998-1G
APOSOR912998-1GEA 386 EUR
APOSOR912998-1G
3-Chloro-5-nitropyridine 95%
3-Chloro-5-nitropyridine
Formel: C₅H₃ClN₂O₂
Molecular Weight: 158,543 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04114218
CAS-Nummer: 22353-33-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR