3-Amino-4-phenylpyridine 95%

Lieferant: Apollo Scientific

Synonyme: 4-Phenylpyridin-3-amine

OR912999-1G
APOSOR912999-1GEA 304 EUR
APOSOR912999-1G
3-Amino-4-phenylpyridine 95%
3-Amino-4-phenylpyridine
Formel: C₁₁H₁₀N₂
Molecular Weight: 170,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04114255
CAS-Nummer: 146140-99-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR