1-Butyl-2-methylindole 95%
Lieferant: Apollo Scientific
OR913042-5G
APOSOR913042-5GEA
108
EUR
APOSOR913042-5G
1-Butyl-2-methylindole 95%
1-Butyl-2-methylindole
Formel:
C₁₃H₁₇N Molecular Weight: 187.281 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04972048 CAS-Nummer: 42951-35-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...