C-(1-Piperidin-1-ylcyclohexyl)methylamine 98%

Lieferant: Apollo Scientific

OR913056-1G
APOSOR913056-1GEA 415 EUR
APOSOR913056-1G
C-(1-Piperidin-1-ylcyclohexyl)methylamine 98%
C-(1-Piperidin-1-ylcyclohexyl)methylamine
Formel: C₁₂H₂₄N₂
Molecular Weight: 196,34 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05201339
CAS-Nummer: 41805-36-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR