2-Oxo-1,2-dihydro-[1,8]naphthyridine-3-carboxylic acid 98%

Lieferant: Apollo Scientific

Synonyme: 2-Oxo-1,2-dihydro-1,8-naphthyridine-3-carboxylic acid

OR913168-1G OR913168-250MG
APOSOR913168-1GEA 212 EUR
APOSOR913168-1G APOSOR913168-250MG
2-Oxo-1,2-dihydro-[1,8]naphthyridine-3-carboxylic acid 98%
2-Oxo-1,2-dihydro-[1,8]naphthyridine-3-carboxylic acid
Formel: C₉H₆N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD06659017
CAS-Nummer: 5175-14-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR