(S)-(-)-N-Methoxymethyl-N-(trimethylsilyl)methyl-1-phenylethylamine

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-Methoxymethyl-(1-phenyl-ethyl)-trimethylsilanylmethyl-amine

OR913190-1G
APOSOR913190-1GEA 284 EUR
APOSOR913190-1G
(S)-(-)-N-Methoxymethyl-N-(trimethylsilyl)methyl-1-phenylethylamine
(S)-(-)-N-Methoxymethyl-N-(trimethylsilyl)methyl-1-phenylethylamine
Formel: C₁₄H₂₅NOSi
Molecular Weight: 251,45 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06798960
CAS-Nummer: 143140-08-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR