2-Methylamino-1-piperidin-1-yl-ethanone hydrochloride 95%

Lieferant: Apollo Scientific

OR913287-1G
APOSOR913287-1GEA 130 EUR
APOSOR913287-1G
2-Methylamino-1-piperidin-1-yl-ethanone hydrochloride 95%
2-Methylamino-1-piperidin-1-yl-ethanone hydrochloride
Formel: C₈H₁₆N₂O·ClH
Molecular Weight: 192,686 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08060487
CAS-Nummer: 1176419-66-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR