1-Acetyl-5-chloro-pyrazolo-[3,4-c]pyridine 95%

Lieferant: Apollo Scientific

Synonyme: 1-Acetyl-5-chloropyrazolo[3,4-c]pyridine

OR913288-1G
APOSOR913288-1GEA 956 EUR
APOSOR913288-1G
1-Acetyl-5-chloro-pyrazolo-[3,4-c]pyridine 95%
1-Acetyl-5-chloro-pyrazolo-[3,4-c]pyridine
Formel: C₈H₆ClN₃O
Molecular Weight: 195,606 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08061149
CAS-Nummer: 76006-04-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR