4-Amino-N-methylindole 97%

Lieferant: Apollo Scientific

Synonyme: 1-Methyl-1H-indol-4-amine

OR913289-1G
APOSOR913289-1GEA 78.3 EUR
APOSOR913289-1G
4-Amino-N-methylindole 97%
4-Amino-N-methylindole
Formel: C₉H₁₀N₂
Molecular Weight: 146,189 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08061153
CAS-Nummer: 85696-95-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR