3-(4-Methylphenyl)-1H-pyrazole 95%

Lieferant: Apollo Scientific

Synonyme: 3-(p-Tolyl)-1H-pyrazole

OR9133-5G
APOSOR9133-5GEA 148 EUR
APOSOR9133-5G
3-(4-Methylphenyl)-1H-pyrazole 95%
3-(4-Methylphenyl)-1H-pyrazole
Formel: C₁₀H₁₀N₂
Molecular Weight: 158,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00665963
CAS-Nummer: 59843-75-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR