2-(Benzo[d]oxazol-2-yl)-2-methylpropanenitrile 95%

Lieferant: Apollo Scientific

OR913301-1G OR913301-250MG
APOSOR913301-1GEA 171 EUR
APOSOR913301-1G APOSOR913301-250MG
2-(Benzo[d]oxazol-2-yl)-2-methylpropanenitrile 95%
2-(Benzo[d]oxazol-2-yl)-2-methylpropanenitrile
Formel: C₁₁H₁₀N₂O
Molecular Weight: 186.2100 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08166396
CAS-Nummer: 157763-81-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR