3-Phenylpentan-3-amine hydrochloride 95%
Lieferant: Apollo Scientific
OR913327-1G
APOSOR913327-1GEA
393
EUR
APOSOR913327-1G
3-Phenylpentan-3-amine hydrochloride 95%
3-Phenylpentan-3-amine hydrochloride
Formel:
C11H18ClN Lagertemperatur: Raumtemperatur |
CAS-Nummer:
104177-96-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...