Methyl 3-pyrrolidinecarboxylate hydrochloride 98%

Lieferant: Apollo Scientific

Synonyme: Methyl pyrrolidine-3-carboxylate hydrochloride

OR913341-25G
APOSOR913341-25GEA 229 EUR
APOSOR913341-25G
Methyl 3-pyrrolidinecarboxylate hydrochloride 98%
Methyl 3-pyrrolidinecarboxylate hydrochloride
Formel: C₆H₁₁NO₂·ClH
Molecular Weight: 165.618 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08436169
CAS-Nummer: 198959-37-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR