N-Boc-3-methylenepiperidine 95%

Lieferant: Apollo Scientific

OR913374-1G
APOSOR913374-1GEA 0 EUR
APOSOR913374-1G
N-Boc-3-methylenepiperidine 95%
N-Boc-3-methylenepiperidine
Formel: C₁₁H₁₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08706141
CAS-Nummer: 276872-89-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR