(2-Acetyl-phenoxy)-acetic acid 95%

Lieferant: Apollo Scientific

OR913438-1G
APOSOR913438-1GEA 142 EUR
APOSOR913438-1G
(2-Acetyl-phenoxy)-acetic acid 95%
(2-Acetyl-phenoxy)-acetic acid
Formel: C₁₀H₁₀O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00118956
CAS-Nummer: 1878-62-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR