(3-Pyridyl)-thiophen-2-ylmethanol 95%

Lieferant: Apollo Scientific

OR913517-1G
APOSOR913517-1GEA 522 EUR
APOSOR913517-1G
(3-Pyridyl)-thiophen-2-ylmethanol 95%
(3-Pyridyl)-thiophen-2-ylmethanol
Formel: C₁₀H₉NOS
Molecular Weight: 191.25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01108848
CAS-Nummer: 21314-77-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR