2,6-Dimethyl-4-nitroanisol 96%

Lieferant: Apollo Scientific

Synonyme: 2-Methoxy-1,3-dimethyl-5-nitrobenzene

OR913588-5G
APOSOR913588-5GEA 183 EUR
APOSOR913588-5G
2,6-Dimethyl-4-nitroanisol 96%
2,6-Dimethyl-4-nitroanisol
Formel: C₉H₁₁NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00024537
CAS-Nummer: 14804-39-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR