(E)-Methyl 3-(2-chlorophenyl)acrylate 98%

Lieferant: Apollo Scientific

OR913589-1G
APOSOR913589-1GEA 296 EUR
APOSOR913589-1G
(E)-Methyl 3-(2-chlorophenyl)acrylate 98%
(E)-Methyl 3-(2-chlorophenyl)acrylate
Formel: C₁₀H₉ClO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09788608
CAS-Nummer: 98288-14-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR