(R)-4-Benzyl-1,3-thiazolidine-2-one 95%
Lieferant: Apollo Scientific
OR913714-1G
APOSOR913714-1GEA
290
EUR
APOSOR913714-1G
(R)-4-Benzyl-1,3-thiazolidine-2-one 95%
(R)-4-Benzyl-1,3-thiazolidine-2-one
Formel:
C₁₀H₁₁NOS Molecular Weight: 193,265 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06796568 CAS-Nummer: 1217647-47-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...