Dimethyl (S)-(-)-methylsuccinate 95%

Lieferant: Apollo Scientific

Synonyme: (S)-(-)-Methylsuccinic Acid Dimethyl Ester, (S)-(-)-Pyrotartaric Acid Dimethyl Ester, Dimethyl (S)-(-)-Pyrotartarate

OR913785-1G
APOSOR913785-1GEA 364 EUR
APOSOR913785-1G
Dimethyl (S)-(-)-methylsuccinate 95%
Dimethyl (S)-(-)-methylsuccinate
Formel: C₇H₁₂O₄
Molecular Weight: 160,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00134572
CAS-Nummer: 63163-08-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR